Labels for BMP™ 51/53 printer, type B-490, Freezerbondz™
Leverancier: Brady
FreezerBondz
BRDY143310EA 73.72 EUR
BRDY143310 BRDY143309 BRDY143314 BRDY143312 BRDY143311
Labels for BMP™ 51/53 printer, type B-490, Freezerbondz™
Etiketten
Freezerbondz™ labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen.
  • They have good print smudge and solvent resistance
  • Good high and low temperature performance
  • For tube/vial applications these labels must be wrapped upon itself with at least a 3 mm overlap