Nikkel(II)sulfaat hexahydraat 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
|
Gevaar
|
2808-05EA
0
EUR
2808-05
2808-01
2808-04
2808-R
Nikkel(II)sulfaat hexahydraat 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Nikkel(II)sulfaat hexahydraat
Formule:
NiSO₄·6H₂O MW: 262,85 g/mol Smeltpunt: 840 °C Dichtheid: 2,07 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00149813 CAS-nummer: 10101-97-0 EINECS: 232-104-9 UN: 3077 ADR: 9,III |
Specificaties testresultaten
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (NiSO₄·6H₂O) (by EDTA titrn) | 98.0 - 102.0 % |
Insoluble Matter | ≤0.005 % |
Chloride (Cl) | ≤0.001 % |
Nitrogen Compounds (as N) | ≤0.002 % |
Calcium (Ca) | ≤0.005 % |
Cobalt (Co) | ≤0.002 % |
Copper (Cu) | ≤0.005 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Magnesium (Mg) | ≤0.005 % |
Manganese (Mn) | ≤0.002 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...