Benzoyl fluoride 97%

Gevaar

ACRO402040050EA 28.5 EUR
ACRO402040050 ACRO402040250
Benzoyl fluoride 97%
Benzoyl fluoride
Formule: C₇H₅FO
Kookpunt: 159…161 °C (760 mmHg)
Smeltpunt: –28 °C
Dichtheid: 1,15 g/cm³
Vlampunt: 48 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000364
CAS-nummer: 455-32-3
UN: 2920
ADR: 8,II

Bestel nu

Specificaties testresultaten

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR