Isoquinoline-3-carboxylic acid 97%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO454490010EA
113
EUR
ACRO454490010
Isoquinoline-3-carboxylic acid 97%
Isoquinoline-3-carboxylic acid
Formule:
C₁₀H₇NO₂ MW: 173,1681 g/mol Smeltpunt: -170…171,5 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00075137 CAS-nummer: 6624-49-3 |
Specificaties testresultaten
Appearance | White to light brown Solid |
Infrared spectrum | Conforms |
Water | ≤10 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...