Lamivudine 98%

Leverancier: Thermo Scientific
Waarschuwing

ACRO461090010EA 139 EUR
ACRO461090010
Lamivudine 98%
Lamivudine
Formule: C₈H₁₁N₃O₃S
MW: 229,2600 g/mol
Smeltpunt: 160…162 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00869739
CAS-nummer: 134678-17-4

Bestel nu

Specificaties testresultaten

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 175 to 179 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR