VWRK610260
Leverancier: Clin-Tech
VWRK610260EA 20.1 EUR
VWRK610260
VWRK610260
A component of Perl’s method to demonstrate the presence of ferric salts and siderotic granules in blood films, bone marrow or histology sections.
See also product code 610270 (Perls’ solution 2)
Bestel nu