Concanamycin A (from Streptomyces species) ≥98% (by HPLC)

Leverancier: Enzo Life Sciences

ALEXALX380034M001EA 2680 EUR
ALEXALX380034M001 ALEXALX380034C025 ALEXALX380034C100
Concanamycin A (from Streptomyces species) ≥98% (by HPLC)
Concanamycin A

White to off-white solid (sub-milligram quantities will appear as clear film)


  • Isolated from Streptomyces sp.


Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1. Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Exhibits cytotoxic effects in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production. Inhibits Autophagy.


Soluble in methanol, DMSO or acetonitrile, insoluble in water


Verpakking: Shipping temperature: Ambient

Formule: C₄₆H₇₅NO₁₄
Opslagtemperatuur: Vriezer
CAS-nummer: 80890-47-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR