Oligomycin B (from Streptomyces sp. MST-AS5339) ≥95% (by HPLC)

Leverancier: Enzo Life Sciences

ALEXALX380038M005EA 292 EUR
ALEXALX380038M005
Oligomycin B (from Streptomyces sp. MST-AS5339) ≥95% (by HPLC)
Oligomycin B

F-ATPase inhibitor.


Minor component of the oligomycin complex. Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1F0), preventing phosphoryl group transfer. Induces apoptosis.


White to off-white powder


Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO


Verpakking: Shipping temperature: Ambient

Formule: C₄₅H₇₂O₁₂
Opslagtemperatuur: Vriezer
CAS-nummer: 11050-94-5
https://be.vwr.com/stibo/web/std.lang.all/04/52/45390452.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR