Concanamycin C (from Streptomyces species) ≥96% (by HPLC)
ALEXALX380099C500EA 2200 EUR
ALEXALX380099C500
Concanamycin C (from Streptomyces species) ≥96% (by HPLC)
Concanamycin C
V-ATPase inhibitor.
Inhibits vacuolar-type H+-ATPase. Potential antiosteoporotic agent. Shows antiviral and immunosuppressive activity. Active against fungi and yeasts.
White to off-white solid
Soluble in chloroform, methanol, acetonitrile or DMSO. Insoluble in water
Verpakking: Shipping temperature: Ambient
Formule: C₄₅H₇₄O₁₃
Opslagtemperatuur: Vriezer
CAS-nummer: 81552-34-3