Echinosporin (from Streptomyces species) ≥98% (by HPLC)
ALEXALX380115M001EA
285
EUR
ALEXALX380115M001
ALEXALX380115M005
Echinosporin (from Streptomyces species) ≥98% (by HPLC)
Echinosporin
Antitumor antibiotic which inhibits cell cycle at the G(2)/M phase and induces apoptosis.
White to off-white solid
Soluble in dimethyl formamide, methanol or water
Verpakking: Shipped on Blue Ice
Formule:
C₁₀H₉NO₅ Opslagtemperatuur: Vriezer |
CAS-nummer:
79127-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...