Hexacyclinic acid (from Streptomyces cellulosae ssp. griseorubinosus) ≥98%

ALEXALX380123M005EA 0 EUR
ALEXALX380123M005
Hexacyclinic acid (from Streptomyces cellulosae ssp. griseorubinosus) ≥98%
Hexacyclinic acid

Antitumor antibiotic


Displays weak cytotoxicity against different tumor cell lines.


White to pale pink solid


Soluble in DMSO, methanol or acetone


Verpakking: Shipping temperature: Ambient

Formule: C₂₆H₃₄O₉
Opslagtemperatuur: Vriezer
CAS-nummer: 309757-85-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR