Antibiotic PF 1052 (from Phoma sp.) ≥98% (by HPLC)

Leverancier: Enzo Life Sciences

ALEXALX380147M001EA 380 EUR
ALEXALX380147M001 ALEXALX380147M005
Antibiotic PF 1052 (from Phoma sp.) ≥98% (by HPLC)
Antibiotic PF 1052

Antibacterial, antifungal and phytotoxic agent.


Light brown solid


Soluble in methylene chloride, methanol and chlorophorm


Verpakking: Shipping temperature: Ambient

Formule: C₂₆H₃₉NO₄
Opslagtemperatuur: Vriezer
CAS-nummer: 147317-15-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR