Malformin C ≥97% (by HPLC)

ALEXALX380318MC25EA 128 EUR
ALEXALX380318MC25
Malformin C ≥97% (by HPLC)
Malformin C

Inhibitor of bleomycin-induced G2 arrest. Has antibacterial, antimalarial and antitrypanosomal activity. Induces root curvature and malformation in plants and enhances fibrinolytic activity in plants.


White solid


Soluble in 100% ethanol, chloroform, DMSO or methanol; insoluble in water


Verpakking: Shipping temperature: Ambient

Formule: C₂₃H₃₉N₅O₅S₂
Opslagtemperatuur: Vriezer
CAS-nummer: 59926-78-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR