Virustomycin A (from Streptomyces sp. AM-2604) ≥97% (by HPLC)

Leverancier: Enzo Life Sciences

ALEXALX380323M005EA 618 EUR
ALEXALX380323M005
Virustomycin A (from Streptomyces sp. AM-2604) ≥97% (by HPLC)
Virustomycin A

18-membered macrolide. Active against trichomonads and various RNA and DNA viruses. Weakly active against some fungi. Has antitrypanosomal activity.


  • Antiviral agent


Yellow solid


Soluble in acetone, chloroform, 100% ethanol or methanol; insoluble in water or hexane


Verpakking: Shipping temperature: Ambient

Formule: C₄₈H₇₁NO₁₄
Opslagtemperatuur: Vriezer
CAS-nummer: 84777-85-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR