Spectinomycin sulphate ≥98% (by HPLC)
Leverancier: Enzo Life Sciences
ALEXBML-A281-0010EA
77
EUR
ALEXBML-A281-0010
Spectinomycin sulphate ≥98% (by HPLC)
Spectinomycin sulphate
Antibiotic. Inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. Effective against Gram-negative and Gram-positive bacteria (Gonnococcus only). Mutation in rpsE (the gene for ribosomal protein S5) prevents binding of spectinomycin.
- White solid
Soluble in water
Verpakking: Shipping temperature: Ambient
Formule:
C₁₄H₂₄N₂O₇·H₂SO₄ Opslagtemperatuur: Vriezer |
MDL-nummer:
MFCD00270185 CAS-nummer: 23312-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...