Pyripyropene A ≥85% (by HPLC)

Leverancier: Enzo Life Sciences

ALEXALX350134C500EA 283 EUR
ALEXALX350134C500
Pyripyropene A ≥85% (by HPLC)
Pyripyropene A

ACAT2 inhibitor


Highly specific inhibitor of acyl-coenzyme A:cholesterol acetyltransferase 2 (ACAT2).


Isolated from Aspergillus fumigatus FO-1289.


Bestelinformatie: Fungal strain courtesy of The Kitasato Institute, Tokyo.

MDL-nummer: MFCD00929109
CAS-nummer: 147444-03-9
https://be.vwr.com/stibo/web/std.lang.all/76/83/45347683.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR