Curvularin ≥95%
ALEXALX380305M005EA 304 EUR
ALEXALX380305M005 ALEXALX380305M001
Curvularin ≥95%
Curvularin
Inhibitor of iNOS expression
Inhibits expression of inducible nitric oxide synthase (iNOS; NOSII). Has antifungal and antibiotic activity. Inhibits cell division. Similar to Curvulin.
CAS-nummer: 10140-70-2