Radicicol ≥97%

Leverancier: Enzo Life Sciences

ALEXBML-EI285-0001EA 159 EUR
ALEXBML-EI285-0001
Radicicol ≥97%
Radicicol

HSP90 inhibitor


Antifungal macrocyclic lactone antibiotic with antimalarial activity. Potent inhibitor of HSP90. Binds more strongly to HSP90 (nanomolar affinity) than to Grp94. Also binds to yeast HSP90, E. coli HtpG and TRAP-1. Non-competitive inhibitor of ATP citrate lyase. Anti-angiogenic. Specifically inhibits the interaction between HIF-1α/Arnt heterodimer and the hypoxia-responsive element (HRE), reducing VEGF expression. Protein tyrosine kinase inhibitor, e.g. p60v-src (IC50=0.27nM). Inhibitor of cyclooxygenase-2 (COX-2) expression (IC50=27nM) without affecting COX-1 expression in LPS-stimulated macrophages. Induces the differentiation of HL-60 cells into macrophages, blocking cell cycle at G1 and G2. Suppressor of NIH 3T3 cell transformation by diverse oncogenes such as mos, ras and src in part by blocking the key signal transduction intermediates such as MAP kinase and GAP-associated p62. Inhibitor of AP-1-, NF-κB- and serum response factor (SRF)-mediated transcription (e.g. expression of iNOS). Represses the transcriptional function of the estrogen receptor. Inhibits archeal growth and DNA topoisomerase VI (a Topo IIB family topoisomerase). Blocks replication of negative-strand RNA viruses.Causes morphological reversion of ras-transformed cells and leads to selective depletion of Raf kinase. Displays anti-angiogenic activity. Binds to and inhibits Hsp 90 molecular chaperone.


Isolated from Humicola fuscoatra.

Formule: C₁₈H₁₇ClO₆
MW: 364,8 g/mol
CAS-nummer: 12772-57-5
https://be.vwr.com/stibo/web/std.lang.all/37/38/45403738.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR