Oleic acid ≥99% (by GC)

Leverancier: Enzo Life Sciences

ALEXBML-FA045-0010EA 105 EUR
ALEXBML-FA045-0010
Oleic acid ≥99% (by GC)
Oleic acid

PKC activator


Major mono-unsaturated fatty acid constituent of olive oil occurring as the glyceride. Activates protein kinase C (PKC) in hepatocytes. Inhibitor of glucose production and food intake. Upregulates the expression of uncoupling protein-3 (UCP-3) in L6 myotubes. Promotes differentiation of cultured neuronal cells mediated via PPARβ and it induces a myofibroblast-like phenotype in mesangial cells.

Formule: C₁₈H₃₄O₂
MW: 282,47 g/mol
MDL-nummer: MFCD00064242
CAS-nummer: 112-80-1
https://be.vwr.com/stibo/web/std.lang.all/89/57/2298957.jpg https://be.vwr.com/stibo/web/std.lang.all/38/56/45403856.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR