Polydimethylsiloxane, 350 cSt

Leverancier: Serva

Synoniemen: Siliconenolie

SERA35136.01EA 47.9 EUR
SERA35136.01
Polydimethylsiloxane, 350 cSt
Polydimethylsiloxane
Dichtheid: 0,966…0.972 g/ml (25 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00084411
CAS-nummer: 63148-62-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR