Aluminium
SIALIRMM523CEA
292
EUR
SIALIRMM523C
Aluminium
Aluminium
Formule:
Al MW: 26,98 g/mol Kookpunt: 2327 °C (1013 hPa) Smeltpunt: 660 °C Dichtheid: 2,7 g/cm³ (20 °C) |
MDL-nummer:
MFCD00134029 CAS-nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...