Indium ≥99.0%, granular, Sigma-Aldrich®
Leverancier: Merck
M-Clarity™ quality level = MQ200
1.12196.0100EA
546
EUR
1.12196.0100
Indium ≥99.0%, granular, Sigma-Aldrich®
Indium
Formule:
In MW: 114,82 g/mol Dichtheid: 7,31 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00134048 CAS-nummer: 7440-74-6 EINECS: 231-180-0 ADR: 4.1,III |
Specificaties testresultaten
Assay (complexometric) | ≥ 99.0 % |
Particle size | about 2 - 8 mm |
Identity (ICP-OES) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...