Carbon ≥99%, felt, Thickness: 2.54 cm (1.0 in)
Leverancier: Thermo Scientific
43201.RFEA
179
EUR
43201.RF
Carbon ≥99%, felt, Thickness: 2.54 cm (1.0 in)
Carbon
Formule:
C MW: 12,01 g/mol Kookpunt: ∼4000 °C (1013 hPa) Smeltpunt: 3727 °C Dichtheid: 3,65 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00133992 CAS-nummer: 7440-44-0 EINECS: 231-955-3 Merck Index: 13,01818 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...