Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Leverancier: Thermo Scientific
|
Gevaar
|
Packaged under argon.
12381.FIEA
477
EUR
12381.FI
Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Dysprosium
Formule:
Dy MW: 162,5 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00010982 CAS-nummer: 7429-91-6 EINECS: 231-073-9 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...