Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.5 mm (0.02 in)

Leverancier: Thermo Scientific

10229.FFEA 683 EUR
10229.FF
Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.5 mm (0.02 in)
Lutetium
Formule: Lu
MW: 174,97 g/mol
Kookpunt: 3000 °C (1013 hPa)
Smeltpunt: 1651…1675 °C
Dichtheid: 9,849 g/cm³ (25 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011098
CAS-nummer: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Bestel nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR