Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Leverancier: Thermo Scientific
Gevaar


Packaged under argon.

10374.FIEA 1250 EUR
10374.FI
Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Thulium
Formule: Tm
MW: 168,93 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011281
CAS-nummer: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1,II

Bestel nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR