Tantalum, sputtering target, Thickness: 6,35 mm

40895.KSEA 1710 EUR
40895.KS
Tantalum, sputtering target, Thickness: 6,35 mm
Tantalum
Formule: Ta
MW: 180,95 g/mol
Kookpunt: 5427 °C (1013 hPa)
Smeltpunt: 2996 °C
Dichtheid: 16,69 g/cm³ (20 °C)
MDL-nummer: MFCD00011252
CAS-nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR