Guanosine 3′,5′-cyclic monophosphate (cGMP) 1 mM

PROMV6411EA 69 EUR
PROMV6411
Guanosine 3′,5′-cyclic monophosphate (cGMP) 1 mM
Guanosine 3′,5′-cyclic monophosphate (cGMP)

cGMP (guanosine-3´,5´-cyclic monophosphate) is an activator of cGMP-Dependent Protein Kinase.


  • Activator of cGMP-Dependent Protein Kinase
  • Supplied at 1mM concentration


Storage: -30°C to -10°C

Formule: C₁₀H₁₂N₅O₇P
MDL-nummer: MFCD00070129
CAS-nummer: 7665-99-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR