Tantalum, powder, Sigma-Aldrich®

Leverancier: Merck
Gevaar


M-Clarity™ quality level = MQ200

1.12349.0025EA 321 EUR
1.12349.0025
Tantalum, powder, Sigma-Aldrich®
Tantalum
Formule: Ta
MW: 180,95 g/mol
Dichtheid: 16,6 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011252
CAS-nummer: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,

Bestel nu

Specificaties testresultaten

Solubility  - 0.010 g/l (20 °C)
Melting Point 2996 °C
Molar Mass 180.94 g/mol
Bulk Density 500 kg/m3
Boiling Point 5425 °C
Density 16.6 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) (aqueous suspension)
Ignition Point 250 °C
Hill Formula Ta

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR