1,3-Cyclohexanediol cis- and trans-mixture 98%

ACRO111140250EA 66.2 EUR
ACRO111140250 ACRO111140100
1,3-Cyclohexanediol cis- and trans-mixture 98%
1,3-Cyclohexanediol
Formule: C₆H₁₂O₂
MW: 116,16 g/mol
Kookpunt: 246 °C (1013 hPa)
Smeltpunt: 30 °C
Vlampunt: >110 °C
MDL-nummer: MFCD00039458
CAS-nummer: 504-01-8
EINECS: 207-979-5

Bestel nu

Specificaties testresultaten

Appearance Clear colorless to light yellow Viscous liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR