Gentamicin sulphate, Calbiochem®
|
Gevaar
|
345814-1EA
130
EUR
345814-1
Gentamicin sulphate, Calbiochem®
Gentamicin sulphate
Smeltpunt:
218…237 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00270181 CAS-nummer: 1405-41-0 EINECS: 215-778-9 Merck Index: 13,04403 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...