Zetmeel

EDQMS1300000EA 234 EUR
EDQMS1300000
Zetmeel
Zetmeel
Formule: (C₆H₁₀O₅)n
MW: 162,14 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00082026
CAS-nummer: 9005-25-8
EINECS: 232-679-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR