Polyethylene glycol 100,000

Leverancier: Serva

SERA33125.02EA 0 EUR
SERA33125.02 SERA33125.03
Polyethylene glycol 100,000
Polyethylene glycol 100,000
Formule: H(OCH₂CH₂)nOH
MW: 100000 g/mol
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 25322-68-3
EINECS: 500-038-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR