di-Sodium L(+)-tartrate dihydrate
Leverancier: LOHMANN
LOHM36262.9025EA
0
EUR
LOHM36262.9025
di-Sodium L(+)-tartrate dihydrate
di-Sodium L(+)-tartrate dihydrate
Formule:
NaOOC(CHOH)₂COONa·2H₂O MW: 230,08 g/mol Smeltpunt: 57 °C Dichtheid: 1,818 g/cm³ (20 °C) |
MDL-nummer:
MFCD00150035 CAS-nummer: 6106-24-7 EINECS: 212-773-3 Merck Index: 12,08833 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...