L(+)-Cysteine hydrochloride monohydrate

Leverancier: BACHEM BIOCHEMICA

BACE4030461.0100EA 209 EUR
BACE4030461.0100 BACE4030461.0250
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate
Formule: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175,64 g/mol
Smeltpunt: 175 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00065606
CAS-nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://be.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR