(±)-2-Mercaptopropionic acid for synthesis, Sigma-Aldrich®
Gevaar
M-Clarity™ quality level = MQ200
8.22087.9999 8.22087.9037 8.22087.9038
(±)-2-Mercaptopropionic acid for synthesis, Sigma-Aldrich®
(±)-2-Mercaptopropionic acid
Formule: C₃H₆O₂S
MW: 106,15 g/mol
Kookpunt: 205 °C (1013 hPa)
Smeltpunt: 10…14 °C
Dichtheid: 1,2 g/cm³ (20 °C)
Vlampunt: 110 °C
MDL-nummer: MFCD00004862
CAS-nummer: 79-42-5
EINECS: 201-206-5
UN: 2936
ADR: 6.1,II
Merck Index: 12,09476
Bestel nu
Specificaties testresultaten
Solubility (20 °C) soluble
Melting Point 10 - 14 °C
Molar Mass 106.14 g/mol
Boiling Point 203 - 208 °C (1013 hPa)
Vapor Pressure 1.7 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4809 (20 °C, 589 nm)
Density 1.20 g/cm3 (20 °C)
pH 2 (H2O, 20 °C) (undiluted)