(±)-2-Methylvaleric acid

Leverancier: EHRENSTORFER GMBH
Gevaar

EHERC15147200EA 54.2 EUR
EHERC15147200
(±)-2-Methylvaleric acid
(±)-2-Methylvaleric acid
Formule: C₆H₁₂O₂
MW: 116,16 g/mol
Kookpunt: 195 °C (1013 hPa)
Smeltpunt: –85 °C
Dichtheid: 0,92 g/cm³ (20 °C)
Vlampunt: 87 °C
MDL-nummer: MFCD00002671
CAS-nummer: 97-61-0
EINECS: 202-594-9
UN: 3265
ADR: 8,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR