(±)-2-Methyl-2,4-pentaandiol 99+%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO219450500EA
26.8
EUR
ACRO219450500
(±)-2-Methyl-2,4-pentaandiol 99+%
(±)-2-Methyl-2,4-pentaandiol
Formule:
C₆H₁₄O₂ MW: 118,18 g/mol Kookpunt: 197 °C (760 mmHg) Smeltpunt: –40 °C Dichtheid: 0,922 g/cm³ Vlampunt: 93 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00004547 CAS-nummer: 107-41-5 EINECS: 203-489-0 Merck Index: 13,04730 |
Specificaties testresultaten
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4260 to 1.4280 (20°C, 589 nm) |
GC | ≥99 % |
Color scale | ≤15 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...