(R)-(-)-1-Phenylethyl isothiocyanate ≥95%
Leverancier: Thermo Scientific
|
Gevaar
|
L20315.03EA
71
EUR
L20315.03
(R)-(-)-1-Phenylethyl isothiocyanate ≥95%
(R)-(-)-1-Phenylethyl isothiocyanate
Formule:
C₉H₉NS MW: 163,24 g/mol Kookpunt: 132…133 °C (20 mmHg) Dichtheid: 1,060 Vlampunt: 103 °C (217 °F) Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00135478 CAS-nummer: 24277-44-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...