Hydroxylammonium sulphate 99%

Waarschuwing

ACRO198531000EA 24.5 EUR
ACRO198531000 ACRO198530010 ACRO198530051
Hydroxylammonium sulphate 99%
Hydroxylammonium sulphate
Formule: H₈N₂O₆S
MW: 164,14 g/mol
Smeltpunt: 170 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00044869
CAS-nummer: 10039-54-0
EINECS: 233-118-8
UN: 2865
ADR: 8,III
Merck Index: 13,04853

Bestel nu

Specificaties testresultaten

Appearance White Crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR