Hydrazinium sulphate
Leverancier: Molekula
|
Gevaar
|
MOLE29155210-1KGEA
0
EUR
MOLE29155210-1KG
Hydrazinium sulphate
Hydrazinium sulphate
Formule:
NH₂NH₂·H₂SO₄ MW: 130,12 g/mol Smeltpunt: 254 °C Dichtheid: 1,37 g/cm³ (20 °C) |
MDL-nummer:
MFCD00044873 CAS-nummer: 10034-93-2 EINECS: 233-110-4 UN: 3288 ADR: 6.1,III Merck Index: 13,04791 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...