Semicarbazide hydrochloride

Leverancier: EHRENSTORFER GMBH
Gevaar

EHERC16933500EA 118 EUR
EHERC16933500
Semicarbazide hydrochloride
Semicarbazide hydrochloride
Formule: CH₆ClN₃O
MW: 111,53 g/mol
Smeltpunt: 174…178 °C
MDL-nummer: MFCD00013009
CAS-nummer: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08588

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR