5-Sulphosalicylic acid dihydrate
Leverancier: Molekula
|
Waarschuwing
|
MOLE27492718-1KGEA
0
EUR
MOLE27492718-1KG
5-Sulphosalicylic acid dihydrate
5-Sulphosalicylic acid dihydrate
Formule:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O MW: 254,22 g/mol Smeltpunt: 224 °C Dichtheid: 1,695 g/cm³ (20 °C) Vlampunt: 150 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00149540 CAS-nummer: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8,III Merck Index: 12,09138 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...