Piperazine hexahydrate
|
Gevaar
|
EDQMP2100000EA
208
EUR
EDQMP2100000
Piperazine hexahydrate
Piperazine hexahydrate
Formule:
C₄H₁₀N₂·6H₂O MW: 194,23 g/mol Kookpunt: 145 °C (1013 hPa) Smeltpunt: 44…45 °C Vlampunt: 87 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00149389 CAS-nummer: 142-63-2 EINECS: 203-808-3 UN: 2579 ADR: 8,III Merck Index: 12,07617 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...