Ammonium iron(II) sulphate hexahydrate 99% ACS, Fluka™
215406-100GEA
39.2
EUR
215406-100G
215406-500G
Ammonium iron(II) sulphate hexahydrate 99% ACS, Fluka™
Ammonium iron(II) sulphate hexahydrate
Formule:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392,14 g/mol Smeltpunt: 100 °C Dichtheid: 1,86 g/cm3 (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00150530 CAS-nummer: 7783-85-9 |
Specificaties testresultaten
Assay (manganometric) | 98.5 - 101.5 % |
Insoluble in H₂SO₄ | Max. 0.01 % |
Calcium (Ca) | Max. 0.005 % |
Copper (Cu) | Max. 0.003 % |
Potassium (K) | Max. 0.002 % |
Magnesium (Mg) | Max. 0.002 % |
Manganese (Mn) | Max. 0.01 % |
Sodium (Na) | Max. 0.02 % |
Zinc (Zn) | Max. 0.003 % |
Iron(III) | Max. 0.01 % |
Phosphate (PO₄) | Max. 0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...