Ytterbium(III) chloride hexahydrate 99.9%

Waarschuwing

ACRO194430100EA 88.2 EUR
ACRO194430100 ACRO194430010 ACRO194430500
Ytterbium(III) chloride hexahydrate 99.9%
Ytterbium(III) chloride hexahydrate
Formule: Cl₃Yb·6H₂O
MW: 387,49 g/mol
Smeltpunt: 865 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149885
CAS-nummer: 10035-01-5
EINECS: 233-800-5

Bestel nu

Specificaties testresultaten

Appearance White Crystalline powder or crystals and/or chunks
Total conc. of imp.as REO ≤0.1 %
Ytterbium (Yb) 43.1 to 46.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR