Copper(II) nitrate hemipentahydrate
|
Gevaar
|
HONE31288.9050
Copper(II) nitrate hemipentahydrate
Copper(II) nitrate hemipentahydrate
Formule:
Cu(NO₃)₂·2.5H₂O MW: 232,59 g/mol Smeltpunt: 255 °C Dichtheid: >1 g/cm³ (20 °C) |
MDL-nummer:
MFCD02253001 CAS-nummer: 19004-19-4 EINECS: 221-838-5 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...