Zinc sulphate monohydrate USP

Leverancier: VWR Chemicals
Gevaar

29246.298EA 0 EUR
29246.298
Zinc sulphate monohydrate USP
Zinc sulphate monohydrate
Formule: ZnSO₄·1H₂O
MW: 179,45 g/mol
Dichtheid: 3,2 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149893
CAS-nummer: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 13,10213

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR