Manganese(II) sulphate tetrahydrate, EMSURE® for analysis, Supelco®

Leverancier: Merck
Gevaar


M-Clarity™ quality level = MQ300

1.02786.1000EA 129 EUR
1.02786.1000 1.02786.9025
Manganese(II) sulphate tetrahydrate, EMSURE® for analysis, Supelco®
Manganese(II) sulphate tetrahydrate
Formule: MnO₄S·4H₂O
MW: 223,06 g/mol
Dichtheid: 2,11 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149160
CAS-nummer: 10101-68-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III

Bestel nu

Specificaties testresultaten

Solubility (20 °C) soluble
Melting Point 26 - 27 °C
Molar Mass 223.06 g/mol
Bulk Density 1000 - 1200 kg/m3
Density 2.11 g/cm3 (20 °C)
pH 4.5 - 6.0 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR