(+)-Dimethyl-L-tartrate

Leverancier: Molekula

MOLEM55649443EA 0 EUR
MOLEM55649443
(+)-Dimethyl-L-tartrate
(+)-Dimethyl-L-tartrate
Formule: C₆H₁₀O₆
MW: 178,14 g/mol
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 58…61 °C
Dichtheid: 1,3403 g/cm³ (15 °C)
MDL-nummer: MFCD00064437
CAS-nummer: 608-68-4
EINECS: 210-166-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR